missing translation for 'onlineSavingsMsg'
Learn More

OHAUS™ Frontier™ 5306 Mini Centrifuge Accessory, Adapters

For use with Frontier 5306 Mini Centrifuge

Brand:  OHAUS™ 30129562

457.42 NOK valid until 2025-12-31
Use promo code "25339" to get your promotional price.


 View more versions of this product

Product Code. 15826622

  • 492.00 NOK / Pack of 8

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

  • 0.2 mL Centrifuge Tube Adapter
  • Holds up to 8 tubes
  • 0.2 mL Centrifuge Tube Adapter
  • Holds up to 8 tubes
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Adapter
Frontier 5306 mini centrifuge
0.5 mL
0/1 V
8 x 0.5 mL
Adapter, 0.5ml Tube x8, FC5306
8 Pieces
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

One moment while we fetch your results.
Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
OHAUS™ Frontier™ 5306 Mini Centrifuge Accessory, Adapters > For 0.5mL tube

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.