Learn More
Fisher Diagnostics™ ThromboScreen™ 200/400C Accessories
Accessories for ThromboScreen 200/400C
Brand:  Fisher Diagnostics™ 106025
Product Code. 11474549
  • 355.00 NOK / 1 L
Specifications
Pipet Tips
ThromboScreen 200/400
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
4 results found
Lot Number Certificate Type Date Catalog Number
Lot Number62525A1 Certificate TypeCertificate of Analysis Date20/05/2025 Catalog Number106025
Lot Number62522M1 Certificate TypeCertificate of Analysis Date07/11/2023 Catalog Number106025
Lot Number62522D1 Certificate TypeCertificate of Analysis Date07/03/2023 Catalog Number106025
Lot Number62520F1 Certificate TypeCertificate of Analysis Date27/07/2021 Catalog Number106025
Promotions
Promotions
Product Content Correction
Product Title
Fisher Diagnostics™ ThromboScreen™ 200/400C Accessories > Pipet tips