Learn More
Thermo Scientific™ Finnpipette™ Stands
GREENER_CHOICE
Provide safe and convenient storage for manual and electronic pipettes using these stands, with options for single stand, F-stand, multi-stand, multichannel and carousel.
Brand:  Thermo Scientific™ 9420360
1088.04 NOK valid until 2025-07-13
Use promo code "24135" to get your promotional price.
 View more versions of this product
Product Code. 11726924
  • 1315.00 NOK / Each
Elevate Your Beverage Testing with Precision Solutions
Use promo code 24135 at checkout
View Available Promotions
Only null left
Estimated Shipment: 17-06-2025
Description
Provide safe and convenient storage for manual and electronic pipettes using Thermo Scientific™ Finnpipette™ Stands. Available options include options for single stand, F-stand, multi-stand, multichannel and carousel.
Finnpipette F-Stand
  • Accommodates six pipettes
  • Sturdy crossbar structure with slot for each pipette for secure storage
  • Suitable for both single channel and multichannel models
Finnpipette Multichannel Stand
  • Multi-position stand
  • Can hold any multichannel Finnpipette
  • Pipette can be stored in upright or horizontal position
  • Small footprint
Finnpipette Carousel Stand
  • Rotating, portable
  • Transparent lid protects pipettes from contamination
  • Shipped with three adapters for electronic Finnpipette Novus pipettes and five adapters for manual Finnpipette pipettes
Finnpipette Novus Stand
  • For convenient storage of electronic Finnpipette Novus pipette
  • Accommodates both single and multichannel Finnpipette Novus models with ease
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Pipette Stand
White
Electronic Pipette
Videos
  • Video Player is loading.
    Current Time 0:00
    Duration 0:00
    Loaded: 0%
    Stream Type LIVE
    Remaining Time 0:00
    1x
    • Chapters
    • descriptions off, selected
    • captions off, selected
      Thermo Scientific Finnpipette Novus electronic pipettors
    • Video Player is loading.
      Current Time 0:00
      Duration 0:00
      Loaded: 0%
      Stream Type LIVE
      Remaining Time 0:00
      1x
      • Chapters
      • descriptions off, selected
      • captions off, selected
        Thermo Scientific Finnpipette Novus electronic pipettors
      Certificates
      Promotions
      Promotions
      Product Content Correction
      Product Title
      Thermo Scientific™ Finnpipette™ Stands > Finnpipette Novus Single Pipette Stand