Learn More
Fisherbrand™ Fiberboard Box Dividers for Ultra-Low Temperature and Cryogenic Freezers
Organize vial storage within cryo boxes with these fiberboard box dividers.
Brand:  Fisherbrand™ 5958
373.78 NOK valid until 2025-06-30
Use promo code "24156" to get your promotional price.
Product Code. 11986924
  • 413.86 NOK / Pack of 12
Spotlight
Use promo code 24156 at checkout
View Available Promotions
Only null left
In Stock
Description
  • Use with fiberboard storage boxes to divide box into grids for vial storage
  • Box dividers for cold storage application available in 7 x 7 , 8 x 8, 9 x 9 or 10 x 10 grids
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Vials (100)
Certificates
Promotions
Promotions
  • Multiple Manufacturers
    Save up to 59% on some of our most popular products
    Promo Code: 24140
    Expires2025-08-24
  • Multiple Manufacturers
    Save big on essential products for cancer research
    Promo Code24147
    Expires2025-07-31
  • Multiple Manufacturers
    Big discounts are available on a range of popular products including chemicals, lab essentials, equipment, consumables, life science and safety products.
    Promo Code: 24156
    Expires2025-06-30
    Hot Deal
    *Discount will apply to qualifying orders received by Fisher Scientific between 01/05/2025 and 30/06/2025. Promotion code must be presented at the time of order. Discount applies to the list price in effect at the time the order is received. Promotion cannot be combined with other discounts or promotions. Offer void where prohibited, licensed, or restricted by federal, state, provincial, or local laws or regulation or agency/institutional policy. Other restrictions may apply.
    Terms & Conditions See Full Promo Details
  • Multiple Manufacturers
    Browse exclusive prices for our academic customers
    Promo Code: 25081
    Expires2025-12-31
Product Content Correction
Product Title
Fisherbrand™ Fiberboard Box Dividers for Ultra-Low Temperature and Cryogenic Freezers >