Learn More
Lonza™ FGM™ Fibroblast Growth Medium BulletKit™
Complete kit, basal medium and supplements, to support growth of fibroblasts.
Brand:  Lonza™ CC-3130
Product Code. 11615430
  • 5400.00 NOK / Each
Explore more special offers
Description
Description
  • Culture system containing FBM™ Basal Medium (CC-3131), and; FGM SingleQuots™ Supplement Pack (CC-4134).
  • Developed to support growth of fibroblasts.
  • Includes the following:
    • 1 x FBM™ Basal Medium (CC-3131), 500 mL
    • 1 x FGM SingleQuots™ Supplement Pack (CC-4134) containing the following:
      • 1 x Lilac Cap Vial with Insulin, 0.50 mL
      • 1 x Gray Cap Vial with hFGF-B, 0.50 mL
      • 1 x Red Cap Vial with GA-1000, 0.50 mL
      • 1 x Bottle FBS, 10.00 mL
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Red
Fibroblast Cells
7.4 to 7.7
Basal Medium, Insulin, hFGF-B, GA-1000, FBS; Store in dark at 2 to 8°C
Basal Medium and Supplements
Medium: 500 mL, Insulin: 0.5 mL, hFGF-B: 0.5 mL, GA-1000: 0.5 mL, FBS: 10.0 mL