missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FGFR1OP2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11605-1-AP-150UL
This item is not returnable.
View return policy
Description
May be involved in wound healing pathway.Specifications
FGFR1OP2 | |
Polyclonal | |
Unconjugated | |
FGFR1OP2 | |
FGFR1 oncogene partner 2, FGFR1OP2, HSPC123 like | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
26127, 362463, 67529 | |
-20°C | |
Liquid |
Western Blot | |
0.17 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q6TA25, Q9CRA9, Q9NVK5 | |
FGFR1OP2 | |
FGFR1OP2 Fusion Protein Ag2153 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction