missing translation for 'onlineSavingsMsg'
Learn More

FGF-10 Antibody (014), FITC, Novus Biologicals™

Rabbit Monoclonal Antibody

Brand:  Novus Biologicals NBP2-89572F

Product Code. 18606310

  • 5805.00 NOK / 0.10mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

FGF-10 Monoclonal antibody specifically detects FGF-10 in Human samples. It is validated for ELISA
TRUSTED_SUSTAINABILITY
Specifications

Specifications

FGF-10
Monoclonal
FITC
PBS
Rabbit
Protein A purified
RUO
Primary
Human
Purified
ELISA
014
ELISA
FGF-10, fibroblast growth factor 10, Keratinocyte growth factor 2, produced by fibroblasts of urinary bladder lamina propria
This antibody was obtained from a rabbit immunized with purified, recombinant Human FGF-10 (NP_004456.1; Gln38-Ser208).
0.1 mL
Angiogenesis, Biologically Active Proteins, Cell Cycle and Replication
2255
Store at 4°C in the dark.
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
FGF-10 Antibody (014), FITC, Novus Biologicals™ > 0.1 mL; FITC

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.