missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ferric Ammonium Sulfate Dodecahydrate, ≥99%, (ACS Reagent Grade), MP Biomedicals
missing translation for 'orderingAttributeHoverText'
Quantity:
500 g
Spezifikation
Spezifikation
CAS | 7783-83-7 |
Quantity | 500 g |
Assay Percent Range | ≥99% |
Molecular Formula | FeH28NO20S2 |
Synonym | ammonium ferric sulfate, ferric ammonium sulfate dodecahydrate, iron ammonium sulfate dodecahydrate, sulfuric acid, ammonium iron 3+ salt 2:1:1 , dodecahydrate, ferric ammonium sulfate, acs, azanium iron 3+ disulfate dodecahydrate, ferric iron ammonium dodecahydrate disulfate, azanium iron +3 cation disulfate dodecahydrate, ammonium iron iii sulfate dodecahydrate, acs 100g |
InChI Key | LCPUDZUWZDSKMX-UHFFFAOYSA-K |
SMILES | [NH4+].O.O.O.O.O.O.O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[Fe+3] |
IUPAC Name | azanium;iron(3+);disulfate;dodecahydrate |
Molecular Weight (g/mol) | 482.176 |
PubChem CID | 197097 |
Mehr anzeigen |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'