Learn More
Thermo Scientific™ DEPC-treated Water
Shop All Thermo Scientific Products
Click to view available options
Quantity:
30 mL
5 x 1 mL
Unit Size:
30mL
Pack of 5
Description
deionized, diethylpyrocarbonate (DEPC) is treated and 0.22 ÎĽm membrane-filtered water. It is ideal for applications involving RNA.
Ideal for applications involving RNA.
- Free of endo-, exodeoxyribonucleases, ribonucleases and phosphatases
Note:
DEPC-treated Water is not recommended for PCR or in vivo experiments (1).
Specifications
Specifications
pH | 7.9 |
Grade | Molecular Biology |
Packaging | Bottle |
Treatment(s) | DEPC-Treated |
Quantity | 30 mL |
Documents
Product Information
Manuals
User Guide: DEPC-treated Water
Version: Jan. 2015
File size: 50.9 kB
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number | Certificate Type | Date | Product Code |
---|---|---|---|
3149297 | Certificate of Analysis | 13/02/2025 | R0601,10213130 |
2811735 | Certificate of Analysis | 13/02/2025 | R0601,10213130 |
3129908 | Certificate of Analysis | 13/12/2024 | R0601,10213130 |
2757543 | Certificate of Analysis | 04/07/2023 | R0601 |
2699543 | Certificate of Analysis | 11/04/2023 | R0601 |
For Research Use Only. Not for use in diagnostic procedures.
Product Content Correction
Product Title
Thermo Scientific™ DEPC-treated Water >