Learn More
Thermo Scientific™ DNA Gel Loading Dye (6X)
GREENER_CHOICE
Prepare DNA markers and samples for loading on agarose or polyacrylamide gels with Thermo Scientific™ 6X DNA Loading Dye.
Brand:  Thermo Scientific™ R0611
274.01 NOK valid until 2025-07-04
Use promo code "24112" to get your promotional price.
Product Code. 11541575
  • 411.00 NOK / Pack of 5
Get 3 for 2
Use promo code 24112 at checkout
View Available Promotions
Alert:
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”24112” to get your promotional price
Description
Description
6X DNA Loading Dye is used to prepare DNA markers and samples for loading on agarose or polyacrylamide gels. It contains two different dyes (bromophenol blue and xylene cyanol FF) for visual tracking of DNA migration during electrophoresis. The presence of glycerol ensures that the DNA in the ladder and sample forms a layer at the bottom of the well. The EDTA included in the solution binds divalent metal ions and inhibits metal-dependent nucleases.
6X DNA Loading Dye is used for conventional DNA electrophoresis.
Highlights
  • Two-color tracking of DNA migration during DNA electrophoresis
  • No DNA masking during gel exposure to UV light
  • EDTA binds divalent metal ions and inhibits metal dependent nucleases
TRUSTED_SUSTAINABILITY
Specifications
Specifications
6 X
DNA Gel Loading Dye
Acrylamide Gels, Agarose Gels
5 x 1.0 mL
SDS
Documents Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number2807160 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog NumberR0611
Lot Number2793408 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog NumberR0611
Lot Number2824996 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog NumberR0611
Lot Number2824942 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog NumberR0611
Lot Number2850777 Certificate TypeCertificate of Analysis Date28/01/2025 Catalog NumberR0611
Promotions
Promotions
Product Content Correction
Product Title
Thermo Scientific™ DNA Gel Loading Dye (6X) > 5 x 1mL