Learn More
Value Plastics Female Luer Thread Style to UNF Threaded Adapter Fittings, Straight
Luer fittings allow for a simple yet secure connection
407.65 NOK - 598.57 NOK
Product Code | Brand | Thread Size | Inlet | Material (Seal) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Thread Size | Inlet | Material (Seal) | Price | Quantity & Availability | ||||
30053983
|
Value Plastics
50120-85 |
1/4-28 UNF(M) | Female Luer with 5/16 in. Hex | Polypropylene |
598.57 NOK
Pack of 25 |
Estimated Shipment: 01-08-2025
Log in to see stock. |
||||
30053201
|
Value Plastics
50120-92 |
1/4-28 UNF(M) | Female Luer with 5/16 in. Hex | Polypropylene |
562.92 NOK
Pack of 25 |
Estimated Shipment: 01-08-2025
Log in to see stock. |
||||
30053683
|
Value Plastics
50120-00 |
10-32 UNF(M) | Female Luer with 1/4 in. Hex | Nylon |
407.65 NOK
Pack of 25 |
Estimated Shipment: 01-08-2025
Log in to see stock. |
||||
30054148
|
Value Plastics
50120-35 |
1/4-28 UNF(M) | Female Luer with 5/16 in. Hex | Nylon |
427.14 NOK
Pack of 25 |
Estimated Shipment: 15-09-2025
Log in to see stock. |
||||
30053992
|
Value Plastics
50120-84 |
1/4-28 UNF(M) | Female Luer with 5/16 in. Hex | Nylon |
407.65 NOK
Pack of 25 |
Estimated Shipment: 15-09-2025
Log in to see stock. |
||||
30054264
|
Value Plastics
50120-86 |
1/4-28 UNF(M) | Female Luer with 5/16 in. Hex | Polycarbonate |
575.91 NOK
Pack of 25 |
Estimated Shipment: 01-08-2025
Log in to see stock. |
||||
30054098
|
Value Plastics
50120-01 |
10-32 UNF(M) | Female Luer with 1/4 in. Hex | Polycarbonate |
575.91 NOK
Pack of 25 |
Estimated Shipment: 01-08-2025
Log in to see stock. |
||||
Description
Quick and easy assembly and disassembly without the need for tools. Thread design provides a secure closure—preventing contamination. Conforms to ISO-594-1 and ISO 594-2 standardsSpecifications
25/Pk. |
Product Title
Value Plastics Female Luer Thread Style to UNF Threaded Adapter Fittings, Straight