Learn More
FBL Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 16021-1-AP-20UL
 View more versions of this product
Product Code. 16895464
  • 1696.32 NOK / 20µL
Explore more special offers
Description
Description
Nop1p / Fibrillarin was originally identified as a nucleolar protein of bakers yeast, Saccharomyces cerevisiae (accession P15646). The Nop1p protein is essential for yeast viability and is localized in the nucleoli. The human homologue of Nop1p is fibrillarin (accession P22087) a component of the nucleolar small nuclear ribonucleoprotein (snRNP) particle. The human fibrillarin gene is located on chromosome 19 (19q13.1).
Specifications
Specifications
FBL
Polyclonal
Unconjugated
FBL
FBL, FIB, FIB1, fibrillarin, FLRN, RNU3IP1
Rabbit
Antigen Affinity Chromatography
RUO
14113, 2091, 292747
-20°C
Liquid
Immunoprecipitation, Western Blot, Immunofluorescence, Immunocytochemistry
0.25 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P22087, P22509, P35550
FBL
FBL Fusion Protein Ag8841
20 μL
Primary
Rat, Mouse, Human