missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals Fatty Acid (Essential) Deficient Diet, Powder

Essential fatty acid deficient diet. Contains stearic acid U.S.P./N.F.

Brand:  MP Biomedicals 0296023950

 View more versions of this product

Product Code. 11437121

  • 36015.00 NOK / 50kg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Essential fatty acid deficient diet
  • Contains Stearic acid U.S.P/N.F.
  • Also contains casein and other vitamins and minerals
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Autoclavable
Single-use
50 kg
Room Temperature
0.3048 m
Animal Diet
Powder
Non-sterile
1 ft.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.