missing translation for 'onlineSavingsMsg'
Learn More

FANCL Mouse anti-Human, Clone: 6E2B6, Proteintech

Mouse Monoclonal Antibody

1696.32 NOK - 4796.50 NOK

Specifications

Antigen FANCL
Clone 6E2B6
Concentration 2 mg/mL
Applications Western Blot
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16848705
View Documents
Proteintech
66639-1-IG-20UL
20 μL
1696.32 NOK
20µL
Please to purchase this item. Need a web account? Register with us today!
16838705
View Documents
Proteintech
66639-1-IG-150UL
150 μL
4796.50 NOK
150µL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

The Fanconi anemia complementation group (FANC) currently includes FANCA, FANCB, FANCC, FANCD1 (also called BRCA2), FANCD2, FANCE, FANCF, FANCG, FANCI, FANCJ (also called BRIP1), FANCL, FANCM and FANCN (also called PALB2). The previously defined group FANCH is the same as FANCA. Fanconi anemia is a genetically heterogeneous recessive disorder characterized by cytogenetic instability, hypersensitivity to DNA crosslinking agents, increased chromosomal breakage, and defective DNA repair. The members of the Fanconi anemia complementation group do not share sequence similarity; they are related by their assembly into a common nuclear protein complex. This gene encodes the protein for complementation group L. Alternative splicing results in two transcript variants encoding different isoforms.
Specifications

Specifications

FANCL
2 mg/mL
Monoclonal
Liquid
RUO
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
FAAP43, FANCL, Fanconi anemia group L protein, PHF9, POG
FANCL
IgG2a
Protein A
Antibody
6E2B6
Western Blot
Unconjugated
Mouse
Human
Q9NW38
55120
FANCL Fusion Protein Ag6192
Primary
-20°C
FANCL
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
FANCL Mouse anti-Human, Clone: 6E2B6, Proteintech

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.