Learn More
Falcon™ Pipet Controller Replacement Parts
Assortment of replacement parts for pipet controllers.
Brand: Falcon™ 357482
116.80 NOK valid until 2025-06-30
Use promo code "24075" to get your promotional price.
This item is not returnable.
View return policy
Description
For use with Falcon pipet controller.
Specifications
O-ring | |
Pipet controller |
Promotions
-
Multiple Manufacturers
The Ultimate Lab Appliances Promotion
Enjoy discounts up to 78% on top-quality lab equipment and upgrade your laboratory with the best appliances at the lowest prices.Promo Code: 24075Expires: 2025-06-30
New Offer Save Up to 78%
Product Content Correction
Product Title
Falcon™ Pipet Controller Replacement Parts > O-ring