Description
- Soft, crystal-clear PVC frame goggles with a wide support area all around the face to enhance comfort.
- Specific channel on the side of the frame to accommodate the temple of prescription frames.
- Model fits well with prescription glasses.
- Special neoprene headband for good chemical resistance and easy cleaning.
- Sealed version to protect against liquid droplets and large dust particles.
- Anti-Fog coating to help reduce lens fogging.
Specifications
Specifications
Certifications/Compliance | ETIM Class 5.0, EC002098, CE Approved , EN 166: 2001 |
Lens Coating | Anti-fog |
Description | Sealed |
Product Type | Safety Goggles |
For Use With (Application) | Construction, food industry |
Frame Color | Clear |
Headband Material | Neoprene |
Ventilation Details | Unventilated |
Material | PVC |
Lens Material | Acetate |
Show More |
Product Content Correction
Product Title
3M™ Fahrenheit™ Unventilated Safety Goggles >