Learn More
Memmert™ Factory Calibration (3 Temperatures) for UN110
Calibration at Memmert factory for three temperature value.
Brand:  Memmert™ UN110D00128
Product Code. 17599755
  • 3870.00 NOK / Each
Explore more special offers
Description
Description
  • For three temperature values: 100°C, 160°C, and 220°C
Specifications
Specifications
UN110 Oven
Measuring point chamber center
Certificate for 3 temperatures
Standard Works Calibration Certificate