missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FABP3/H-FABP Rabbit anti-Mouse, Rat, Clone: 9B9D7, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Bio-Techne NBP3-16508-100UL
This item is not returnable.
View return policy
Description
FABP3/H-FABP Monoclonal antibody specifically detects FABP3/H-FABP in Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence
Specifications
FABP3/H-FABP | |
Monoclonal | |
Unconjugated | |
PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Mouse, Rat | |
Purified |
Western Blot, Immunohistochemistry, Immunofluorescence | |
9B9D7 | |
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:50 - 1:200 | |
FABP11, fatty acid binding protein 11, fatty acid binding protein 3, muscle and heart (mammary-derived growthinhibitor), Fatty acid-binding protein 3, Fatty acid-binding protein 3, muscle, fatty acid-binding protein, heart, Heart-type fatty acid-binding protein, H-FABPM-FABP, Mammary-derived growth inhibitor, MDGI, Muscle fatty acid-binding protein, O-FABP | |
A synthetic peptide corresponding to a sequence within amino acids 1-100 of human FABP3/H-FABP (P05413). | |
100 ÎĽg | |
Cell Cycle and Replication, Cholesterol Metabolism, Lipid and Metabolism | |
2170 | |
Store at -20°C. Avoid freeze-thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
FABP3/H-FABP Rabbit anti-Mouse, Rat, Clone: 9B9D7, Novus Biologicals™ > 100 μg; Unconjugated
Spot an opportunity for improvement?Share a Content Correction