missing translation for 'onlineSavingsMsg'
Learn More
Learn More
F(ab')2 Fragment IgG(H+L) Donkey anti-Mouse, R-PE, Secondary Antibody, Proteintech
Donkey anti-Mouse Affinipure Secondary Antibody
Brand: Proteintech SA00008-9-500UL
This item is not returnable.
View return policy
Specifications
F(ab')2 Fragment IgG(H+L) | |
R-PE | |
Donkey | |
500 μL | |
Mouse | |
Liquid |
Immunofluorescence, Flow Cytometry | |
0.01M Sodium phosphate, 0.25M NaCl, 50% glycerol, pH7.6, with 3 mg/mL BSA | |
The antibody was purified from antisera by immunoaffinity chromatography using antigens coupled to agarose beads. | |
Secondary | |
Store at -20°C. Stable for one year after shipment. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction