Learn More
Fisherbrand™ Eyepiece Pair for Steddy Microscopes
2415.00 NOK - 3995.00 NOK
Specifications
Type Eyepiece
For Use With (Equipment) Steddy Microscopes
View More Specs
Products 3
Product Code Brand Magnification Power Price Quantity & Availability  
Product Code Brand Magnification Power Price Quantity & Availability  
12944451
View Documents
Fisherbrand™
7217.1002
10X
2800.00 NOK
Pack of 2
Estimated Shipment: 18-03-2025
to see stock.
Add to basket
12944361
View Documents
Fisherbrand™
7217.1507
15X
3995.00 NOK
Each
Estimated Shipment: 18-03-2025
to see stock.
Add to basket
12934361
View Documents
Fisherbrand™
7217.0506
5X
2415.00 NOK
Each
Estimated Shipment: 28-03-2025
Description
Trinocular configuration allows for the addition of a camera for digital imaging or video camera.
The diamond shaped base provides stability and hand support whilst samples are manipulated on the stage. The zoom ratio of 1x to 4x gives a magnification of 10x to 40x with the standard 10x eyepieces; both with dioptre compensation. Additional eyepieces and corrective lenses allow for a total magnification range from 2.5 to 120x. Both light sources have variable intensity control. The unit comes with frosted glass stage, a blue filter and a heat filter for the incident light source to prevent overheating of the sample.
Robust stereo zoom microscope
  • Integral single piece column stand eliminates vibration and image distortion
  • Ergonomic diamond shaped base
  • Total magnification range 2.5x to 120x
  • Variable intensity transmitted and incident light sources
Specifications
Specifications
Eyepiece
Steddy Microscopes