Learn More
Lenz Laborglasintrumente Extractors Acc. to Twisselmann
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
6185.00 NOK - 7140.00 NOK
Product Code | Brand | Standard Taper Joint Bottom | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Standard Taper Joint Bottom | Price | Quantity & Availability | ||||||
10223911
|
Lenz Laborglasintrumente
05380025 |
NS 29/32 |
6185.00 NOK
Each |
Estimated Shipment: 03-10-2025
Log in to see stock. |
||||||
10283951
|
Lenz Laborglasintrumente
05380031 |
NS 34/35 |
6415.00 NOK
Each |
Estimated Shipment: 03-10-2025
Log in to see stock. |
||||||
10375971
|
Lenz Laborglasintrumente
05380037 |
NS 45/40 |
7140.00 NOK
Each |
Estimated Shipment: 03-10-2025
Log in to see stock. |
||||||
Description
- Extractor head with vacuum jacket
- Made of DURAN™ tubing
- Twisselmann condenser (improved) with thread GL 14
- Round bottom flask
Specifications
Duran Glass |
Product Title
Lenz Laborglasintrumente Extractors Acc. to Twisselmann