Learn More
Lenz Laborglasintrumente Extractor Heads Acc. To Twisselmann
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
1595.00 NOK - 2160.00 NOK
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | ||||||
10335691
|
Lenz Laborglasintrumente
05415131 |
70 mL |
1595.00 NOK
Each |
Estimated Shipment: 27-05-2025
Log in to see stock. |
||||||
10421093
|
Lenz Laborglasintrumente
05415143 |
150 mL |
2160.00 NOK
Each |
Estimated Shipment: 27-05-2025
Log in to see stock. |
||||||
10602862
|
Lenz Laborglasintrumente
05415137 |
100 mL |
2025.00 NOK
Each |
Estimated Shipment: 27-05-2025
Log in to see stock. |
||||||
Description
- Extractor heads
- Acc. to Twisselmann
- With vacuum jacket
- Made of DURAN™ tubing
Specifications
Borosilicate Glass 3.3 | |
1 Ea. |
Product Title
Lenz Laborglasintrumente Extractor Heads Acc. To Twisselmann