Learn More
Lenz Laborglasintrumente Extractor Heads Acc. to Thielepape
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
Brand:  Lenz Laborglasintrumente 05405143
 View more versions of this product
Product Code. 10040241
  • 2595.00 NOK / Each
Only null left
Estimated Shipment: 14-07-2025
Description
  • Extractor heads acc. to Thielepape
  • With NS 2-way stopcock
  • Made of DURAN™ tubing
Specifications
Specifications
150 mL
Borosilicate Glass 3.3
NS 45/40
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Lenz Laborglasintrumente Extractor Heads Acc. to Thielepape > 150 mL