Learn More
Lenz Laborglasintrumente Extractor Heads Acc. to Soxhlet
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
Brand:  Lenz Laborglasintrumente 05395152
 View more versions of this product
Product Code. 10663511
  • 1755.00 NOK / Each
Only null left
Estimated Shipment: 21-08-2025
Description
  • Extractor heads acc. to DIN 12602
  • With cone NS 29/32
  • Available with/without NS stopcock
  • Made of DURAN™ tubing
Specifications
Specifications
300 mL
Borosilicate Glass 3.3
NS 60/46
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Lenz Laborglasintrumente Extractor Heads Acc. to Soxhlet > 300 mL