Learn More
Koettermann™ Exploris™ Tall Storage Cabinet
Click to view available options
Color:
White
Material:
Steel (1.0330)
Unit Size:
Each
Description
- Constructed of high-grade steel (1.0330), electrogalvanised, polyurethane coated
- Soundproofed, double skin doors
Specifications
Specifications
Product Type | Tall Storage Cabinet |
Color | White |
Dimensions (D x W x H) Interior | 50 x 90 x 192 cm |
No. of Doors | 2 |
No. of Shelves | 3 |
Surface Coating | Polyurethane |
Material | Steel (1.0330) |
Product Content Correction
Product Title
Koettermann™ Exploris™ Tall Storage Cabinet >