Learn More
Novus Biologicals™ Exosome Standards (HT29 cell line)
Product Code. 18623800 Shop All Bio Techne Products
6 x 100 μg Vials, Pack of 6
Click to view available options
Quantity:
2 x 100 μg Vials
2 x 30 μg Vials
6 x 100 μg Vials
6 x 30 μg Vials
Unit Size:
Pack of 2
Pack of 6
Highly pure, lyophilized exosome standards with superior stability, optimal for multiple applications
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage Store at 4°C. After reconstitution store at -70°C.
Concentration LYOPH
Quantity 6 x 100 μg Vials
Product Content Correction
Product Title
Novus Biologicals™ Exosome Standards (HT29 cell line) >