Learn More
Novus Biologicals™ Exosome Standards (HEK293 cell line)
Highly pure, lyophilized exosome standards with superior stability, optimal for multiple applications
Brand: Novus Biologicals™ NBP3-11684-60ug
This item is not returnable.
View return policy
Description
Specifications
Store at 4°C. After reconstitution store at -70°C. | |
LYOPH | |
2 x 30 ÎĽg Vials |
Product Content Correction
Product Title
Novus Biologicals™ Exosome Standards (HEK293 cell line) > 2 x 30ug Vials