missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EXOSC3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 15062-1-AP-20UL
This item is not returnable.
View return policy
Description
EXOSC3 is component of the exosome 3′->5′ exoribonuclease complex and is required for the 3′ processing of the 7S pre-RNA to the mature 5.8S rRNA.Specifications
EXOSC3 | |
Polyclonal | |
Unconjugated | |
EXOSC3 | |
CGI 102, EXOSC3, exosome component 3, hRrp 40, hRrp40p, p10, RP11 3J10.8, RRP40, Rrp40p | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
313243, 51010, 66362 | |
-20°C | |
Liquid |
Western Blot, Immunohistochemistry (Paraffin), Immunofluorescence, Immunocytochemistry, Immunoprecipitation | |
0.19 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q7TQK4, Q9NQT5 | |
Exosc3 | |
EXOSC3 Fusion Protein Ag7065 | |
20 μL | |
Primary | |
Mouse, Human, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction