Learn More
EVOS™ EVOS™ Vessel Holder, two 60-mm Petri dishes
For flasks, trays and petri dishes
Brand:  EVOS™ AMEPVH003
Product Code. 11987315
  • 1304.00 NOK / Each
Estimated Shipment: 30-01-2025
to see stock.
Explore more special offers
Description
Broad range of holders helps maximize the flexibility of the EVOS and Micromaster™ inverted microscopes.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Vessel Holder
EVOS™ XL Core Imaging System, EVOS™ FL Color Imaging System, EVOS™ XL Imaging System, EVOS™ FL Imaging System, EVOS™ FL Auto Imaging System
2 60 mm Petri Dishes
EVOS
two, 60 mm Petri dishes