Learn More
Thermo Scientific™ Evolution™ Specular Reflectance Accessories
Get high performance and accuracy for the measurement of glossy or mirrored surfaces at fixed angles with Thermo Scientific™ Evolution™ Specular Reflectance Accessories.
Brand:  Thermo Scientific™ 222-233700
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 11833664
  • 25920.00 NOK / Each
Estimated Shipment: 25-07-2024
Description
Measure at 8, 15, 20, 30, 45 and 60 degrees, or at any angle from 30 degrees to 80 degrees, with Thermo Scientific™ Evolution™ Specular Reflectance Accessories.
This suite of accessories is useful for measuring film thickness and reflectance properties from coatings on metallic, glass, and silicon substrates. The accessory includes an alignment mirror that can also be used for baseline measurements.
Convenient Horizontal Sample Mounting
  • Convenient horizontal sample mounting allows easy positioning of samples for measurement
  • Selection of apertures limits size of the light patch striking the sample surface, facilitating reproducible sampling
  • Align it once and it's aligned for life- no need to align each time you insert the accessory
Versatility with Polarizers in SSH
Can be used in conjunction with a variety of polarizers, depolarizers, and filter holders
Evolution 200/300 Solid Sample Holder accommodates multiple slide holders, allowing optical elements to be inserted either before or after the Fixed-Angle Specular Reflectance accessory to begin your measurements.
Software for QA/QC Testing
Designed for applications where testing at specific angles is required
Ensure testing requirements are met
Specify a table of values or minimum and/or maximum boundaries across an entire spectrum and provide tolerances - the software does the rest
Test results display high and low test boundaries
Text-based confirmation that material fails or exceeds specified limits provided
- See more at: http://www.thermoscientific.com/en/product/fixed-angle-specular-reflectance-accessories.html#sthash.KEtr1HCr.dpuf
Specifications
Specifications
60° Specular Reflectance AccessoryRequires: Sample Slide Holder/Baseplate
Thermo Scientific Evolution One/One Plus and Evolution Pro Spectrophotometers
4 x 13 mm