Learn More
Trajan Scientific and Medical™ eVol™ XR Syringes
Digitally controlled positive displacement dispensing system.
Brand: Trajan Scientific and Medical™ 2910335
This item is not returnable.
View return policy
Description
- Liquid handling user independent device
- Reduction in need for re-analysis
- Syringes easily and quickly changed
- Replacement parts available
Specifications
With GT Plunger | |
eVol Syringe | |
3/Pk. | |
1 mL |
Product Content Correction
Product Title
Trajan Scientific and Medical™ eVol™ XR Syringes > Volume: 1 mL