Learn More
Evocalcet, MedChemExpress Product Code.: 16479897
Evocalcet, MedChemExpress
Product Code. 16479897
5 mg, 5mg
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
Product Code. 16479897
Brand: Medchem Express HY17613
MedChemExpress Evocalcet has an activating effect on calcium sensing receptor (CaSR) extracted from patent WO 2017061621 A1, compound A.
Evocalcet has an activating effect on calcium sensing receptor (CaSR) extracted from patent WO 2017061621 A1, compound A.
Specifications
Chemical Name or Material Evocalcet
CAS 870964-67-3
Color Off-White
Purity Grade Notes Research
Molecular Formula C24H26N2O2
Quantity 5 mg
Synonym KHK7580
Solubility Information DMSO : 30 mg/mL (80.11 mM; Need ultrasonic and warming)
SMILES O=C(O)CC1=CC=C(N2C[C@@H](N[C@@H](C3=C4C=CC=CC4=CC=C3)C)CC2)C=C1
Molecular Weight (g/mol) 374.48
Formula Weight 374.48
Percent Purity 99.05%
Grade Research
Physical Form Solid
For Use With (Application) Metabolism-protein/nucleotide metabolism
Show More Show Less
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Evocalcet, MedChemExpress