Learn More
Thermo Scientific™ Eutech TN-100 Turbidimeter Calibration Kit
Simple, display-prompted push-button calibration
Brand: Thermo Scientific™ 01X621001
3243.02 NOK valid until 2025-06-30
Use promo code "24156" to get your promotional price.
Description
- 60mL bottles of U.S. EPA-approved, non-formazin standards
- Stable and easy to use
Specifications
Calibration Kit Set | |
0.02 NTU, 20.0 NTU, 100 NTU, 800 NTU | |
Standard solutions; 4 vials of 60mL each | |
U.S. EPA-approved non-formazin standards |
Promotions
-
Multiple Manufacturers
The Academic Best Buy Book 2025
Browse exclusive prices for our academic customersPromo Code: 25081Expires: 2025-12-31
-
Multiple Manufacturers
Spotlight
Big discounts are available on a range of popular products including chemicals, lab essentials, equipment, consumables, life science and safety products.Promo Code: 24156Expires: 2025-06-30