missing translation for 'onlineSavingsMsg'
Learn More
Ethyl 5-bromo-1,2,3-thiadiazole-4-carboxylate, 98%, Thermo Scientific Chemicals Product Code.: 11329989

Ethyl 5-bromo-1,2,3-thiadiazole-4-carboxylate, 98%, Thermo Scientific Chemicals

Product Code. 11329989
250 mg
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy

Product Code. 11329989

Brand: Thermo Scientific Alfa Aesar H25764.MD

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Ethyl 5-bromo-1,2,3-thiadiazole-4-carboxylate is used in the preparation of fused 1,3,6- thiadiazepines such as di[1,2,3]triazolo[1,5-a:5',1'-d][3,1,5]benzothiadiazepines.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl 5-bromo-1,2,3-thiadiazole-4-carboxylate is used in the preparation of fused 1,3,6- thiadiazepines such as di[1,2,3]triazolo[1,5-a:5′,1′-d][3,1,5]benzothiadiazepines.

Solubility
Miscible with dimethyl formamide.

Notes
Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6439-91-4
Molecular Formula C5H5BrN2O2S
Molecular Weight (g/mol) 237.071
MDL Number MFCD02324685
InChI Key MBEUYMRLYKXBGT-UHFFFAOYSA-N
Synonym ethyl 5-bromo-1,2,3-thiadiazole-4-carboxylate, 5-bromo-1,2,3-thiadiazole-4-carboxylic acid ethyl ester, acmc-209wgk, 1,2,3-thiadiazole-4-carboxylicacid, 5-bromo-, ethyl ester, 5-bromo-1,2,3-thiadiazole-4-carboxylic acid ethylShow MoreShow Less
PubChem CID 11020903
IUPAC Name ethyl 5-bromothiadiazole-4-carboxylate
SMILES CCOC(=O)C1=C(SN=N1)Br

Specifications

Melting Point 76°C to 78°C
Quantity 250 mg
Solubility Information Miscible with dimethyl formamide.
Formula Weight 237.07
Percent Purity 98%
Chemical Name or Material Ethyl 5-bromo-1,2,3-thiadiazole-4-carboxylate

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

2 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberL03Y011 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberH25764.03,H25764.MD
Lot Number10182983 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberH25764.03,H25764.MD

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Ethyl 5-bromo-1,2,3-thiadiazole-4-carboxylate, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.