missing translation for 'onlineSavingsMsg'
Learn More
Ethyl 4-methyl-2-(3-pyridyl)thiazole-5-carboxylate, 97%, Thermo Scientific™ Product Code.: 15451796

Ethyl 4-methyl-2-(3-pyridyl)thiazole-5-carboxylate, 97%, Thermo Scientific™

Product Code. 15451796
250 mg
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy

Product Code. 15451796

Brand: Thermo Scientific Alfa Aesar H54942.MD

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Ethyl 4-methyl-2-(3-pyridyl)thiazole-5-carboxylate is used as a catalytic agent and petrochemical additive.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 39091-00-4
Molecular Formula C12H12N2O2S
Molecular Weight (g/mol) 248.3
MDL Number MFCD01567252
InChI Key PNJYSPUXYJWHJJ-UHFFFAOYSA-N
Synonym ethyl 4-methyl-2-pyridin-3-yl thiazole-5-carboxylate, ethyl 4-methyl-2-3-pyridinyl-1,3-thiazole-5-carboxylate, ethyl 4-methyl-2-pyridin-3-yl-1,3-thiazole-5-carboxylate, 4-methyl-2-pyridin-3-yl-thiazole-5-carboxylic acid ethyl ester, ethylmethylpyridinylthiazolecarboxylate, ethyl 4-methyl-2-3-pyridyl-5-thiazolecarboxylate, ethyl 4-methyl-2-3-pyridyl thiazole-5-carboxylate, ethyl 4-methyl-2-3-pyridyl-1,3-thiazole-5-carboxylate, ethyl 4-methyl-2-3-pyridinyl-1,3-thiazole-5-carboxylate #, 5-thiazolecarboxylic acid, 4-methyl-2-3-pyridinyl-, ethyl ester
PubChem CID 619088
IUPAC Name ethyl 4-methyl-2-pyridin-3-yl-1,3-thiazole-5-carboxylate
SMILES CCOC(=O)C1=C(N=C(S1)C2=CN=CC=C2)C

Specifications

Quantity 250 mg
Formula Weight 248.3
Percent Purity 97%
Chemical Name or Material Ethyl 4-methyl-2-(3-pyridyl)thiazole-5-carboxylate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Ethyl 4-methyl-2-(3-pyridyl)thiazole-5-carboxylate, 97%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.