missing translation for 'onlineSavingsMsg'
Learn More
Ethyl 4-chloro-5,7-difluoroquinoline-3-carboxylate, 99%, Thermo Scientific™ Product Code.: 11355599

Ethyl 4-chloro-5,7-difluoroquinoline-3-carboxylate, 99%, Thermo Scientific™

Product Code. 11355599
250 mg
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy

Product Code. 11355599

Brand: Thermo Scientific Alfa Aesar H50514.MD

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 311346-69-7
Molecular Formula C12H8ClF2NO2
Molecular Weight (g/mol) 271.648
MDL Number MFCD00173416
InChI Key IEGYWXRVOVIAMV-UHFFFAOYSA-N
Synonym ethyl 4-chloro-5,7-difluoro-3-quinolinecarboxylate, 4-chloro-5,7-difluorolquinoline-3-carboxylic acid ethyl ester
PubChem CID 3743835
IUPAC Name ethyl 4-chloro-5,7-difluoroquinoline-3-carboxylate
SMILES CCOC(=O)C1=C(C2=C(C=C(C=C2N=C1)F)F)Cl

Specifications

Quantity 250 mg
Formula Weight 271.65
Percent Purity 99%
Chemical Name or Material Ethyl 4-chloro-5,7-difluoroquinoline-3-carboxylate

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

3 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10170716 Certificate TypeCertificate of Analysis Date06/12/2021 Catalog Number
Lot NumberD14U031 Certificate TypeCertificate of Analysis Date06/12/2021 Catalog Number
Lot NumberJ13Q030 Certificate TypeCertificate of Analysis Date06/12/2021 Catalog Number
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Ethyl 4-chloro-5,7-difluoroquinoline-3-carboxylate, 99%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.