missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 3-(trifluoromethyl)benzoate, 98%, Thermo Scientific™

Product Code. 11443498
5 g, 5g
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Chemical Identifiers

CAS 76783-59-0
Molecular Formula C10H9F3O2
Molecular Weight (g/mol) 218.175
MDL Number MFCD00013560
InChI Key MHNBTKIAHHECCQ-UHFFFAOYSA-N
Synonym ethyl 3-trifluoromethyl benzoate, 3-trifluoromethyl benzoic acid ethyl ester, ethyl m-trifluoromethylbenzoate, ethyl-3-trifluoromethyl benzoate, ethyl 3-trifluoromethyl-benzoate, benzoic acid, 3-trifluoromethyl-, ethyl ester, ksc496o4j, ethyl a,a,a-trifluoro-m-toluate, 3-trifluoromethylbenzoic acid, ethyl ester
PubChem CID 144790
IUPAC Name ethyl 3-(trifluoromethyl)benzoate
SMILES CCOC(=O)C1=CC(=CC=C1)C(F)(F)F

Specifications

Boiling Point 100°C to 101°C (10 mmHg)
Refractive Index 1.446
Quantity 5 g
Beilstein 1968226
Formula Weight 218.18
Percent Purity 98%
Chemical Name or Material Ethyl 3-(trifluoromethyl)benzoate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Ethyl 3-(trifluoromethyl)benzoate, 98%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.