Learn More
Ethyl 3-methyl-4-nitrobenzoate, 98%, Thermo Scientific™
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications
Specifications
Melting Point | 53°C to 55°C |
Quantity | 50 g |
Synonym | benzoic acid, 3-methyl-4-nitro-, ethyl ester, 3-methyl-4-nitrobenzoic acid ethyl ester, acmc-209zoi, ethyl 3-methyl-4-nitro-benzoate, benzoic acid,3-methyl-4-nitro-, ethyl ester |
Formula Weight | 209.2 |
Percent Purity | 98% |
Chemical Name or Material | Ethyl 3-methyl-4-nitrobenzoate |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Product Code |
---|---|---|---|
10099233 | Certificate of Analysis | 07/12/2021 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.