missing translation for 'onlineSavingsMsg'
Learn More
Ethyl 3-chloro-5-(trifluoromethyl)pyridine-2-carboxylate, 96%, Thermo Scientific Chemicals Product Code.: 15488715

Ethyl 3-chloro-5-(trifluoromethyl)pyridine-2-carboxylate, 96%, Thermo Scientific Chemicals

Product Code. p-7032487
1 g, 1g
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
1g
5g
This item is not returnable. View return policy

Product Code. 15488715

Brand: Thermo Scientific Alfa Aesar H33967.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 128073-16-5
Molecular Formula C9H7ClF3NO2
Molecular Weight (g/mol) 253.605
MDL Number MFCD06656414
InChI Key COMQYNZHBCNPNW-UHFFFAOYSA-N
Synonym ethyl 3-chloro-5-trifluoromethyl picolinate, ethyl 3-chloro-5-trifluoromethyl pyridine-2-carboxylate, ethyl 3-chloro-5-trifluoromethyl-2-pyridinecarboxylate, 2-pyridinecarboxylic acid, 3-chloro-5-trifluoromethyl-, ethyl ester, 3-chloro-5-trifluoromethylpyridine-2-carboxylic acid ethyl ester, ethyl 5-trifluoromethyl-3-chloropyridine-2-carboxylate, ethyl 3-chloro-5-trifluoromethyl picolite, ethyl 3-chloro-5trifluoromethyl-2-pyridinecarboxylate
PubChem CID 22013558
IUPAC Name ethyl 3-chloro-5-(trifluoromethyl)pyridine-2-carboxylate
SMILES CCOC(=O)C1=C(C=C(C=N1)C(F)(F)F)Cl

Specifications

Boiling Point 64°C to 66°C (0.008 mmHg)
Quantity 1 g
Formula Weight 253.6
Percent Purity 96%
Chemical Name or Material Ethyl 3-chloro-5-(trifluoromethyl)pyridine-2-carboxylate

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

2 results found
Lot Number Certificate Type Date Product Code
Lot Number10178233 Certificate TypeCertificate of Analysis Date07/12/2021 Product CodeH33967.03,H33967.09,H33967.06
Lot Number10202419 Certificate TypeCertificate of Analysis Date07/12/2021 Product CodeH33967.03,H33967.09,H33967.06

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Ethyl 3-chloro-5-(trifluoromethyl)pyridine-2-carboxylate, 96%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.