missing translation for 'onlineSavingsMsg'
Learn More
Ethyl 3,5-bis(trifluoromethyl)benzoate, 99%, Thermo Scientific™ Product Code.: 11476826

Ethyl 3,5-bis(trifluoromethyl)benzoate, 99%, Thermo Scientific™

Product Code. 11476826
25 g, 25g
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 11476826

Brand: Thermo Scientific Alfa Aesar B23952.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 96617-71-9
Molecular Formula C11H8F6O2
Molecular Weight (g/mol) 286.17
MDL Number MFCD01320684
InChI Key NBWZJMOEFTYMOF-UHFFFAOYSA-N
Synonym ethyl 3,5-bis trifluoromethyl benzoate, 3,5-bis-trifluoromethyl-benzoic acid ethyl ester, 3,5-bis trifluoromethyl benzoic acid ethyl ester, rarechem al bi 0207, acmc-20al07, ethyl 3?5-bis trifluoromethyl benzoate, ethyl 3 5-bis trifluoromethyl benzoate, benzoic acid, 3,5-bis trifluoromethyl-, ethyl esterShow MoreShow Less
PubChem CID 2733458
IUPAC Name ethyl 3,5-bis(trifluoromethyl)benzoate
SMILES CCOC(=O)C1=CC(=CC(=C1)C(F)(F)F)C(F)(F)F

Specifications

Quantity 25 g
Formula Weight 286.17
Percent Purity 99%
Chemical Name or Material Ethyl 3,5-bis(trifluoromethyl)benzoate

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

1 results found
Lot Number Certificate Type Date Product Code
Lot NumberG7146A Certificate TypeCertificate of Analysis Date07/12/2021 Product Code
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Ethyl 3,5-bis(trifluoromethyl)benzoate, 99%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.