Learn More
Ethyl 2,4-dimethylnicotinate, 96%, Thermo Scientific Chemicals
Description
It is employed in the design, synthesis, and evaluation of a family of propargyl pyridinyl ethers as potential cytochrome p450 inhibitors.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
It is employed in the design, synthesis, and evaluation of a family of propargyl pyridinyl ethers as potential cytochrome p450 inhibitors.
Solubility
Slightly soluble in water.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

Specifications
Specifications
Boiling Point | 116°C to 118°C (10 mmHg) |
Refractive Index | 1.498 |
Quantity | 1 g |
Beilstein | 133910 |
Solubility Information | Slightly soluble in water. |
Formula Weight | 179.22 |
Percent Purity | 96% |
Chemical Name or Material | Ethyl 2,4-dimethylnicotinate |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
10157729 | Certificate of Analysis | 08/12/2021 | L08672.06,L08672.03 |
10170869 | Certificate of Analysis | 08/12/2021 | L08672.06,L08672.03 |
USLY001100 | Certificate of Analysis | 07/12/2021 | L08672.06,L08672.03 |
10159340 | Certificate of Analysis | 07/12/2021 | L08672.06,L08672.03 |
10159463 | Certificate of Analysis | 06/12/2021 | L08672.06,L08672.03 |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.