Learn More
Ethyl 1-(trifluoromethyl)cyclobutanecarboxylate, 97%, Thermo Scientific Chemicals
Product Code. 15488215
Click to view available options
Quantity:
250 mg
1 g
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 1040683-08-6
Molecular Formula C8H11F3O2
Molecular Weight (g/mol) 196.169
MDL Number MFCD10699116
InChI Key AYUISFJYNKIHDD-UHFFFAOYSA-N
PubChem CID 28309116
IUPAC Name ethyl 1-(trifluoromethyl)cyclobutane-1-carboxylate
SMILES CCOC(=O)C1(CCC1)C(F)(F)F
Specifications
Boiling Point 61°C to 63°C (20 mmHg)
Quantity 250 mg
Synonym ethyl 1-trifluoromethyl cyclobutanecarboxylate, 1-trifluoromethyl cyclobutanecarboxylic acid ethyl ester, ethyl 1-trifluoromethyl cyclobutane-1-carboxylate, ethyl1-trifluoromethyl cyclobutanecarboxylate, 1-trifluoromethyl cyclobutanecarboxylic acidethyl ester
Formula Weight 196.17
Percent Purity 97%
Chemical Name or Material Ethyl 1-(trifluoromethyl)cyclobutanecarboxylate