missing translation for 'onlineSavingsMsg'
Learn More
Ethanol, denatured, special, Thermo Scientific™ Product Code.: 15421717

Ethanol, denatured, special, Thermo Scientific™

Product Code. 15421717
1 L
Click to view available options
Quantity:
1 L
4 L
This item is not returnable. View return policy
Regulatory: This product is subject to excise duty on alcohol in accordance with European Directive 92/83/EEC, all customers must have an authorization before delivery.

Product Code. 15421717

Brand: Thermo Scientific Alfa Aesar 041603.K2

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Regulatory: This product is subject to excise duty on alcohol in accordance with European Directive 92/83/EEC, all customers must have an authorization before delivery.

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 64-17-5, 67-56-1, 141-78-6
Molecular Formula C2H6O
Molecular Weight (g/mol) 46.069
MDL Number MFCD00003568
InChI Key LFQSCWFLJHTTHZ-UHFFFAOYSA-N
Synonym ethyl alcohol, alcohol, methylcarbinol, grain alcohol, ethyl hydroxide, ethyl hydrate, algrain, alcohol, anhydrol, tecsol
PubChem CID 702
ChEBI CHEBI:16236
IUPAC Name ethanol
SMILES CCO

Specifications

Color Colorless
Appearance Colorless liquid
Density 0.7893g/mL
Quantity 1 L
Flash Point 13°C (55°F)
Odor Alcohol-like
Physical Form Liquid
Refractive Index 1.361
Beilstein 1718733
Solubility Information Miscible in water.
Formula Weight 46.08
Concentration or Composition (by Analyte or Components) Ethyl alcohol: 92%; Methanol: 4%; Ethyl acetate: 4%
Chemical Name or Material Ethanol
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Ethanol, denatured, special, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.