missing translation for 'onlineSavingsMsg'
Learn More

Ethanol, 96% v/v, Pharmpur™, Scharlab

Product Code. 11384064
Click to view available options
Quantity:
1 L
5 L
This item is not returnable. View return policy
Regulatory: This product is subject to excise duty on alcohol in accordance with European Directive 92/83/EEC, all customers must have an authorization before delivery.
This item is not returnable. View return policy
Regulatory: This product is subject to excise duty on alcohol in accordance with European Directive 92/83/EEC, all customers must have an authorization before delivery.

Used in solvents, as a disinfectant, for pharmaceutical use, in the synthesis of organic products, and in perfumery.

  • Synonyms: Ethyl alcohol, Methylcarbinol, Spirit, Spirit of wine
  • Molecular Formula: C2H5OH
  • Molar Mass: 46.07g/mol
  • CAS: 64-17-5
  • EIN ECS No.: 200-578-6
  • Dielectric Constant: 24.3 at 25°C

Reacts explosively with strong oxidizing agents including chromium trioxide, hydrogen peroxide and inorganic acids such as nitric acid. Can react violently with metals such as magnesium, sodium and potassium to produce hydrogen. May damage gaskets, painted surfaces, protective and sealing grease coatings, materials, rubber and some plastics.

Chemical Identifiers

CAS 64-17-5
Molecular Formula C2H6O
Molecular Weight (g/mol) 46.069
InChI Key LFQSCWFLJHTTHZ-UHFFFAOYSA-N
Synonym ethyl alcohol, alcohol, methylcarbinol, grain alcohol, ethyl hydroxide, ethyl hydrate, algrain, alcohol, anhydrol, tecsol
PubChem CID 702
ChEBI CHEBI:16236
IUPAC Name ethanol
SMILES CCO

Specifications

Melting Point -117°C
Absorbance 0.40 (5.0cm, 240nm)
Acidity 0.003% max (as CH3COOH)
Appearance Clear, Colorless
Density 0.81g/cm3
Boiling Point 78°C
Quantity 1 L
Flash Point 9°C
Identification Passes Test
Ignition Point 425°C
Physical Form Liquid
Residue after Evaporation 0.001% max
Vapor Pressure ∼59hPa at 20°C
Solubility Information Miscible in water at 20°C
Formula Weight 46.07g/mol
Percent Purity 95.1 to 96.9% v/v, 92.6 to 95.2% w/w
Show More Show Less
compliance-icons
Product Identifier
  • Ethanol 96%; extra pure, Pharmpur, Ph Eur, BP
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P240-Ground/bond container and receiving equipment.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P501-Dispose of contents/container to .
Supplemental information
  • MIXTURE LIST-Contains : ethanol
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.