missing translation for 'onlineSavingsMsg'
Learn More
Ethanol 96% +MEK 25 L, Solveco Product Code.: 11365175
compliance-icons

Ethanol 96% +MEK 25 L, Solveco

Product Code. 11365175
25 L, HDPE Drum
Click to view available options
Quantity:
25 L
Packaging:
HDPE Drum
This item is not returnable. View return policy
Regulatory: This product is subject to excise duty on alcohol in accordance with European Directive 92/83/EEC, all customers must have an authorization before delivery.

Product Code. 11365175

Brand: Solveco Chemicals 1010

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product is subject to excise duty on alcohol in accordance with European Directive 92/83/EEC, all customers must have an authorization before delivery.

Fermented ethanol with a concentration of 96%, denatured with methyl ethyl ketone. Technical grade.

Fermented ethanol denatured with methylethylketone. The ethanol has a natural origin and is part of the natural cycle. Solveco Ethanol 96% +MEK is used extensively in the engineering industry and in washing fluids, as well as a coolant in machining.

Specifications

CAS 64-17-5
Color Colorless
Density 0.8109–0.8119kg/L
Boiling Point 78°C
Quantity 25 L
Packaging HDPE Drum
Molecular Formula C2H6O
Synonym ethyl alcohol, alcohol, methylcarbinol, grain alcohol, ethyl hydroxide, ethyl hydrate, algrain, alcohol, anhydrol, tecsol
InChI Key LFQSCWFLJHTTHZ-UHFFFAOYSA-N
SMILES CCO
IUPAC Name ethanol
Molecular Weight (g/mol) 46.069
PubChem CID 702
ChEBI CHEBI:16236
Formula Weight 46.07g/mol
Percent Purity 96%
Grade Technical Grade
Chemical Name or Material Ethyl Alcohol
Show More Show Less
compliance-icons
Product Identifier
  • Denaturerad ETANOL 95% (denaturerad med Metyletylketon)
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P243-Take precautionary measures against static discharge.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.
Supplemental information
  • MIXTURE LIST-Contains : Etanol, Butanon
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Ethanol 96% +MEK 25 L, Solveco >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.