missing translation for 'onlineSavingsMsg'
Learn More

Esculin hydrate, 97%

CAS: 531-75-9 | C15H16O9 | 340.28 g/mol

656.00 NOK - 3435.00 NOK

Chemical Identifiers

CAS 531-75-9
Molecular Formula C15H16O9
Molecular Weight (g/mol) 340.28
MDL Number MFCD00149492
InChI Key XHCADAYNFIFUHF-TYKRLAFXNA-N
Synonym esculin, aesculin, esculoside, --esculin, polychrome, 6,7-dihydroxycoumarin-6-o-glucoside, aesculinum, esculine, esculetin 6-o-glucoside, 6,7-dihydroxycoumarin 6-glucoside
PubChem CID 5281417
ChEBI CHEBI:4853
IUPAC Name 7-hydroxy-6-[(2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxychromen-2-one
SMILES OC[C@H]1O[C@@H](OC2=C(O)C=C3OC(=O)C=CC3=C2)[C@H](O)[C@@H](O)[C@@H]1O
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10146463
SDS View Documents
Thermo Scientific Acros
117830050
5 g Glass bottle
656.00 NOK
5g
Please to purchase this item. Need a web account? Register with us today!
10254430
SDS View Documents
Thermo Scientific Acros
117830100
10 g Glass bottle
1000.00 NOK
10g
Please to purchase this item. Need a web account? Register with us today!
10006253
SDS View Documents
Thermo Scientific Acros
117830500
50 g Glass bottle
3435.00 NOK
50g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Show More
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 208-517-5

Product Identifier
  • Esculin hydrate

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Esculin hydrate, 97%

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.