Learn More
Ertugliflozin, MedChemExpress Product Code.: 16429127
Ertugliflozin, MedChemExpress
Product Code. 16429127
5 mg, 5mg
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
Product Code. 16429127
Brand: Medchem Express HY15461
MedChemExpress Ertugliflozin (PF-04971729) is a potent, selective and orally active inhibitor of the sodium-dependent glucose cotransporter 2 (SGLT2), with an IC50 of 0.877 nM for h-SGLT2. Has the potential for the treatment of type 2 diabetes mellitus.
Ertugliflozin (PF-04971729) is a potent, selective and orally active inhibitor of the sodium-dependent glucose cotransporter 2 (SGLT2), with an IC50 of 0.877 nM for h-SGLT2. Has the potential for the treatment of type 2 diabetes mellitus.
Specifications
Chemical Name or Material Ertugliflozin
CAS 1210344-57-2
Color White
Purity Grade Notes Research
Molecular Formula C22H25ClO7
Quantity 5 mg
Synonym PF-04971729
Solubility Information DMSO : 250 mg/mL (572.24 mM; Need ultrasonic)
SMILES ClC1=CC=C([C@]23O[C@@](CO)(CO3)[C@@H](O)[C@H](O)[C@H]2O)C=C1CC4=CC=C(OCC)C=C4
Molecular Weight (g/mol) 436.88
Formula Weight 436.88
Percent Purity 98.82%
Grade Research
Physical Form Solid
For Use With (Application) Metabolism-sugar/lipid metabolism
Show More Show Less
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Ertugliflozin, MedChemExpress