Learn More
ERp57/ERp60 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 15967-1-AP-150UL
 View more versions of this product
Product Code. 16845384
  • 4796.50 NOK / 150µL
Explore more special offers
Description
Description
This gene encodes a protein of the endoplasmic reticulum that interacts with lectin chaperones calreticulin and calnexin to modulate folding of newly synthesized glycoproteins. The protein was once thought to be a phospholipase; however, it has been demonstrated that the protein actually has protein disulfide isomerase activity. It is thought that complexes of lectins and this protein mediate protein folding by promoting formation of disulfide bonds in their glycoprotein substrates.
Specifications
Specifications
ERp57/ERp60
Polyclonal
Unconjugated
PDIA3
58 kDa microsomal protein, Disulfide isomerase ER 60, ER protein 57, ER protein 60, ER60, ERp57, ERp60, ERp61, GRP57, GRP58, HsT17083, P58, PDIA3, PI PLC, Protein disulfide isomerase A3
Rabbit
Antigen Affinity Chromatography
RUO
14827, 2923, 29468
-20°C
Liquid
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation
0.17 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P11598, P27773, P30101
PDIA3
ERp57/ERp60 Fusion Protein Ag8741
150 μL
Primary
Human, Mouse, Rat