Learn More
Lenz Laborglasinstrumente™ Erlenmeyer Flasks with Conical Ground Joint
For the safe and easy mixing, pouring and storage of chemicals in the laboratory. Lenz Laborglasinstrumente™ Erlenmeyer Flasks with Conical Ground Joint are manufactured from durable DURAN™ borosilicate glass.
Brand:  Lenz Laborglasinstrumente™ 03022428
 View more versions of this product
Product Code. 10064250
  • 1010.00 NOK / Pack of 10
Only null left
Estimated Shipment: 14-07-2025
Description
  • Acc. to DIN EN ISO 4797
  • With scale (approx. volume/mL)
  • With inscription label
  • Corrosion-free and resistant to thermal shock
  • With conical joint, featuring high taper accuracy and excellent tightness
Specifications
Specifications
DIN EN ISO 4797
50 mL
24/29 NS
Conical Ground Joint, Inscription Label
Borosilicate Glass, Duran Glass
51 mm
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Lenz Laborglasinstrumente™ Erlenmeyer Flasks with Conical Ground Joint > Capacity: 50mL; 24/29 NS Socket