missing translation for 'onlineSavingsMsg'
Learn More

ERGIC3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

1696.32 NOK - 4796.50 NOK

Specifications

Antigen ERGIC3
Concentration 0.28 mg/mL
Applications Western Blot, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16855484
View Documents
Proteintech
16029-1-AP-20UL
20 μL
1696.32 NOK
20µL
Please to purchase this item. Need a web account? Register with us today!
16845484
View Documents
Proteintech
16029-1-AP-150UL
150 μL
4796.50 NOK
150µL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

The MW of this protein is 50 kDa, and this antibody specifically recognizes the 50 kDa protein.

ERGIC3 is possible role in transport between endoplasmic reticulum and Golgi.
Specifications

Specifications

ERGIC3
Western Blot, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Unconjugated
Rabbit
Human, Rat, Mouse
Q9CQE7, Q9Y282
296306, 51614, 66366
ERGIC3 Fusion Protein Ag8893
Primary
-20°C
ERGIC3
0.28 mg/mL
Polyclonal
Liquid
RUO
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
C20orf47, CGI 54, ERGIC and Golgi 3, ERGIC3, Erv46, NY BR 84, PRO0989, SDBCAG84
ERGIC3
IgG
Antigen Affinity Chromatography
Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.